missing translation for 'onlineSavingsMsg'
Learn More

3-(Morpholinomethyl)benzaldehyde, 95%, Thermo Scientific™

Product Code. 10629954
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10629954 250 mg 250mg
10595953 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10629954 Supplier Thermo Scientific Maybridge Supplier No. CC40104CB
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 446866-83-7
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
InChI Key KMYNYFKOOXFQGB-UHFFFAOYSA-N
Synonym 3-morpholinomethyl benzaldehyde, 3-morpholin-4-ylmethyl benzaldehyde, benzaldehyde,3-4-morpholinylmethyl, 3-morpholin-4-ylmethyl-benzaldehyde, 3-4-morpholinylmethyl benzaldehyde, 3-morpholinomethylbenzaldehyde, 3-morpholin-4-yl methyl benzaldehyde
PubChem CID 7164603
IUPAC Name 3-(morpholin-4-ylmethyl)benzaldehyde
SMILES C1COCCN1CC2=CC=CC(=C2)C=O

Specifications

Assay Percent Range 95%
Quantity 250 mg
Formula Weight 205.26
Percent Purity 95%
Chemical Name or Material 3-(Morpholinomethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.