| CAS |
612069-31-5
|
| Formula Weight |
888.5017 |
| InChI Formula |
InChI=1S/C44H76N2O14S/c1-15-33-44(11,52)37(48)29(7)45(12)23-25(3)21-42(9,51)39(27(5)36(28(6)40(50)58-33)59-34-22-43(10,55-14)38(49)30(8)57-34)60-41-35(47)32(20-26(4)56-41)46(13)61(53,54)31-18-16-24(2)17-19-31/h16-19,25-30,32-39,41,47-49,51-52H,15,20-23H2,1-14H3/t25-,26-,27+,28-,29-,30+,32+,33-,34+,35-,36+,37-,38+,39-,41+,42-,43-,44-/m1/s1 |
| IUPAC Name |
N-[(2S,3R,4S,6R)-2-[[(2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-2-ethyl-3,4,10-trihydroxy-13-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,6,8,10,12,14-heptamethyl-15-oxo-1-oxa-6-azacyclopentadec-11-yl]oxy]-3-hydroxy-6-methyloxan-4-yl]-N,4-dimethylbenzenesulfonamide |
| Molecular Formula |
C44 H76 N2 O14 S |
| Molecular Weight (g/mol) |
889.14 |
| Percent Purity |
>95% |
| SMILES |
CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)S(=O)(=O)c4ccc(C)cc4)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O |
| Synonym |
1-Oxa-6-azacyclopentadecan-15-one, 13-[(2,6-dideoxy-3-C-methyl-3-O-methyl-alpha-L-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10-trihydroxy-3,5,6,8,10,12,14-heptamethyl-11-[[3,4,6-trideoxy-3-[methyl[(4-methylphenyl)sulfonyl]amino]-beta-D-xylo-hexopyranosyl]oxy]-, (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-, (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-13-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-alpha-L-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10-trihydroxy-3,5,6,8,10,12,14-heptamethyl-11-[[3,4,6-trideoxy-3-[methyl[(4-methylphenyl)sulfonyl]amino]-beta-D-xylo-hexopyranosyl]oxy]-1-oxa-6-azacyclopentadecan-15-one, 3'-N-Demethyl-3'-N-[(4-methylphenyl)sulfonyl]azithromycin, Azithromycin Imp. G (EP), 3'-N-Desmethyl-3'-N-tosyl Azithromycin, Arithromycin Impurity G |
| Chemical Name or Material |
3'-N-Desmethyl-3'-N-tosyl Azithromycin |
| Grade |
HPLC |
|
Show More
Show Less
|
|