missing translation for 'onlineSavingsMsg'
Learn More

3-n-Propylphenol, 98%

Product Code. 11414807
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11414807

Brand: Thermo Scientific Alfa Aesar B21266.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 621-27-2 | C9H12O | 136.194 g/mol

3-n-Propylphenol, is used to produce 2,4,5,6-tetrabromo-3-propylphenol. This reaction will need reagent bromine, aluminum powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-n-Propylphenol, is used to produce 2,4,5,6-tetrabromo-3-propylphenol. This reaction will need reagent bromine, aluminum powder.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-27-2
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD01632130
InChI Key MPWGZBWDLMDIHO-UHFFFAOYSA-N
Synonym 3-n-propylphenol, m-propylphenol, phenol, 3-propyl, phenol, m-propyl, unii-593r721cic, 1-hydroxy-3-n-propylbenzene, phenol, m-propyl-8ci, phenol, 3-propyl-9ci
PubChem CID 69302
IUPAC Name 3-propylphenol
SMILES CCCC1=CC(=CC=C1)O

Specifications

Boiling Point 229°C to 231°C
Refractive Index 1.525
Quantity 5 g
UN Number UN3145
Solubility Information Soluble in water.
Formula Weight 136.19
Percent Purity 98%
Chemical Name or Material 3-n-Propylphenol
compliance-icons compliance-icons
Product Identifier
  • 3-n-Propylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : m-Propylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.