missing translation for 'onlineSavingsMsg'
Learn More

3-n-Propylphenol, 98%

Product Code. 11424807
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424807 25 g 25g
11414807 5 g 5g
11434807 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424807 Supplier Thermo Scientific Alfa Aesar Supplier No. B21266.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 621-27-2 | C9H12O | 136.194 g/mol

3-n-Propylphenol, is used to produce 2,4,5,6-tetrabromo-3-propylphenol. This reaction will need reagent bromine, aluminum powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-n-Propylphenol, is used to produce 2,4,5,6-tetrabromo-3-propylphenol. This reaction will need reagent bromine, aluminum powder.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-27-2
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD01632130
InChI Key MPWGZBWDLMDIHO-UHFFFAOYSA-N
Synonym 3-n-propylphenol, m-propylphenol, phenol, 3-propyl, phenol, m-propyl, unii-593r721cic, 1-hydroxy-3-n-propylbenzene, phenol, m-propyl-8ci, phenol, 3-propyl-9ci
PubChem CID 69302
IUPAC Name 3-propylphenol
SMILES CCCC1=CC(=CC=C1)O

Specifications

Boiling Point 229°C to 231°C
Refractive Index 1.525
Quantity 25 g
UN Number UN3145
Solubility Information Soluble in water.
Formula Weight 136.19
Percent Purity 98%
Chemical Name or Material 3-n-Propylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.