missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-L-tyrosine, MedChemExpress

Product Code. 16495566
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495566 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16495566 Supplier Medchem Express Supplier No. HY113248
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3-Nitro-L-tyrosine is a biomarker of nitrogen free radical species modified proteins in systemic autoimmunogenic conditions.

3-Nitro-L-tyrosine is a biomarker of nitrogen free radical species modified proteins in systemic autoimmunogenic conditions.

Specifications

Chemical Name or Material 3-Nitro-L-tyrosine
CAS 621-44-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C9H10N2O5
Quantity 100 mg
Solubility Information H2O : 5 mg/mL (22.11 mM; ultrasonic and warming and adjust pH to 2 with HCl and heat to 60°C)
SMILES N[C@@H](CC1=CC([N+]([O-])=O)=C(C=C1)O)C(O)=O
Molecular Weight (g/mol) 226.19
Formula Weight 226.19
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.