missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzaldehyde, 99%

Product Code. 10123660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10123660 100 g Plastic bottle 100g
10051440 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10123660 Supplier Thermo Scientific Acros Supplier No. 128411000
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99-61-6 | C7H5NO3 | 151.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-61-6
Molecular Formula C7H5NO3
Molecular Weight (g/mol) 151.12
MDL Number MFCD00007249
InChI Key ZETIVVHRRQLWFW-UHFFFAOYSA-N
Synonym m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde
PubChem CID 7449
IUPAC Name 3-nitrobenzaldehyde
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 56.0°C to 59.0°C
Color Brown-Yellow to Yellow
Boiling Point 285.0°C to 290.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula O2NC6H4CHO
Quantity 100 g
Beilstein 07, 250
Merck Index 14, 6587
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, chloroformm and ether
Formula Weight 151.12
Percent Purity 99%
Physical Form Granular Powder
Chemical Name or Material 3-Nitrobenzaldehyde, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Nitrobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.