missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzaldehyde, 99%

Product Code. 11480134
Click to view available options
Quantity:
250 g
500 g
1000 g
Unit Size:
1000g
250g
500g
This item is not returnable. View return policy

Product Code. 11480134

Brand: Thermo Scientific Alfa Aesar A13594.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-61-6 | C7H5NO3 | 151.121 g/mol

3-Nitrobenzaldehyde is used in the synthesis of other organic compounds, ranging from pharmaceuticals to plastic additives and intermediate for the processing of perfume and flavoring compounds and in the preparation of certain aniline dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrobenzaldehyde is used in the synthesis of other organic compounds, ranging from pharmaceuticals to plastic additives and intermediate for the processing of perfume and flavoring compounds and in the preparation of certain aniline dyes.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-61-6
Molecular Formula C7H5NO3
MDL Number MFCD00007249
InChI Key ZETIVVHRRQLWFW-UHFFFAOYSA-N
Synonym m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde
PubChem CID 7449
IUPAC Name 3-nitrobenzaldehyde

Specifications

Melting Point 57°C to 59°C
Boiling Point 164°C (23 mmHg)
Quantity 250 g
Beilstein 386795
Sensitivity Air Sensitive
Merck Index 14,6587
Solubility Information Slightly soluble in water.
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C=O
Molecular Weight (g/mol) 151.121
Formula Weight 151.12
Percent Purity 99%
Chemical Name or Material 3-Nitrobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.