missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzenesulfonamide, 99%, Thermo Scientific™

Product Code. 15442198
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442198 25 g 25g
15422198 1 g 1g
15432198 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442198 Supplier Thermo Scientific Alfa Aesar Supplier No. H55831.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-52-8
Molecular Formula C6H6N2O4S
Molecular Weight (g/mol) 202.184
MDL Number MFCD00007935
InChI Key TXTQURPQLVHJRE-UHFFFAOYSA-N
Synonym benzenesulfonamide, 3-nitro, m-nitrobenzenesulfonamide, 3-nitrobenzolesulfamide, 3-nitrobenzenesulphonamide, 3-nitrobenzene-1-sulfonamide, m-nitrobenzenesulphonamide, 3-nitro-benzenesulfonamide, benzenesulfonamide, m-nitro, 3-nitrobenzene sulfonamide, m-nitrobenzolsulfonamid
PubChem CID 67138
IUPAC Name 3-nitrobenzenesulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)[N+](=O)[O-]

Specifications

Melting Point 166°C to 168°C
Color White
Quantity 25 g
Formula Weight 202.19
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Nitrobenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.