missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzylamine hydrochloride, 97%

Product Code. 10752611 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10752611 5 g Glass bottle 5g
10347700 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10752611 Supplier Thermo Scientific Acros Supplier No. 184730050
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. Consulta la politica di reso

CAS: 26177-43-5 | C7H8N2O2·HCl | 188.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 26177-43-5
Molecular Formula C7H8N2O2·HCl
Molecular Weight (g/mol) 188.62
MDL Number MFCD00012858
InChI Key DLZXLCHQWOZGSE-UHFFFAOYSA-N
Synonym 3-nitrobenzylamine hydrochloride, 3-nitrophenyl methanamine hydrochloride, 3-nitrobenzylamine hcl, 3-nitrobenzylammonium hydrochloride, benzenemethanamine, 3-nitro-, monohydrochloride, 1-3-nitrophenyl methanamine hydrochloride, pubchem7485, chembl13978, m-nitrobenzylamine hydrochloride, 3-nitrobenzyl amine hydrochloride
PubChem CID 2724030
IUPAC Name (3-nitrophenyl)methanamine;hydrochloride
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl

Specifica

CAS Min % 96.0
CAS Max % 100.0
Melting Point 226°C to 231°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula O2NC6H4CH2NH2·HCl
Quantity 5 g
Beilstein 12, 1076
Formula Weight 188.62
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Nitrobenzylamine hydrochloride, 97%
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • 3-Nitrobenzylamine hydrochloride
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.