missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenol, 98+%

Product Code. 11418196
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418196 10 g 10g
11428196 50 g 50g
11438196 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418196 Supplier Thermo Scientific Alfa Aesar Supplier No. A15312.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 554-84-7 | C6H5NO3 | 139.11 g/mol

3-Nitrophenol is used as indicator in 0.3% Soln in 50% alcohol ph: 6.8 Colorless, 8.6 Yellow. It is used in the synthesis of RAGE inhibitors via the etherification of phenols. Also used in the synthesis of kinase inhibitors against epidermal growth factor receptor T790M mutation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrophenol is used as indicator in 0.3% Soln in 50% alcohol ph: 6.8 Colorless, 8.6 Yellow. It is used in the synthesis of RAGE inhibitors via the etherification of phenols. Also used in the synthesis of kinase inhibitors against epidermal growth factor receptor T790M mutation.

Solubility
Soluble in water (30 mg/ml), ether, alcohol, acetone, and ethanol (700 mg/ml).

Notes
Store in a tightly closed container. Keep from contact with oxidizing materials. Store in a cool, dry, well-ventilated area away from incompatible substances.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 554-84-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007240
InChI Key RTZZCYNQPHTPPL-UHFFFAOYSA-N
Synonym m-nitrophenol, 3-hydroxynitrobenzene, m-hydroxynitrobenzene, phenol, 3-nitro, phenol, m-nitro, m-nitrofenol, meta-nitrophenol, 3-nitro-phenol, m-nitrofenol czech, 1-hydroxy-3-nitrobenzene
PubChem CID 11137
ChEBI CHEBI:34346
IUPAC Name 3-nitrophenol
SMILES OC1=CC=CC(=C1)[N+]([O-])=O

Specifica

Melting Point 93°C to 98°C
Density 1.49
Boiling Point 194°C (70 mmHg)
Flash Point >100°C (212°F)
Quantity 10 g
UN Number UN1663
Beilstein 1907946
Merck Index 14,6619
Solubility Information Soluble in water (30mg/ml),ether,alcohol,acetone,and ethanol (700mg/ml).
Formula Weight 139.11
Percent Purity ≥98%
Chemical Name or Material 3-Nitrophenol
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • 3-Nitrophenol
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Indicazioni di pericolo
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Informazioni supplementari
  • MIXTURE LIST-Contain: m-Nitrophenol

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.