missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenol, 99%

Product Code. 10531431 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10531431

Brand: Thermo Scientific Acros 172300500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 554-84-7 | C6H5NO3 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-84-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007240
InChI Key RTZZCYNQPHTPPL-UHFFFAOYSA-N
Synonym m-nitrophenol, 3-hydroxynitrobenzene, m-hydroxynitrobenzene, phenol, 3-nitro, phenol, m-nitro, m-nitrofenol, meta-nitrophenol, 3-nitro-phenol, m-nitrofenol czech, 1-hydroxy-3-nitrobenzene
PubChem CID 11137
ChEBI CHEBI:34346
IUPAC Name 3-nitrophenol
SMILES OC1=CC=CC(=C1)[N+]([O-])=O

Specifications

Melting Point 94.0°C to 98.0°C
Color Yellow to Brown
Density 1.4900g/mL
Boiling Point 194.0°C (70.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4OH
Quantity 50 g
Beilstein 06,222
Merck Index 15,6706
Specific Gravity 1.49
Solubility Information Solubility in water: 13.5g/L (25°C). Other solubilities: soluble 700g/L in ethanol (20°C),soluble in benzene,diethyl ether,alcohol,,hot and diluted solutions,insoluble in petroleum ehter
Formula Weight 139.11
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrophenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Nitrophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.