missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalic acid, 95%

Product Code. p-4510380 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10235730

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 603-11-2 | C8H5NO6 | 211.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-11-2
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.13
InChI Key KFIRODWJCYBBHY-UHFFFAOYSA-N
Synonym 3-nitrophthalic acid, 3-nitrobenzene-1,2-dicarboxylic acid, phthalic acid, 3-nitro, 1,2-benzenedicarboxylic acid, 3-nitro, 3-nitro phthalic acid, 3-nitro-phthalic acid, 3-nitrophthalicacid, nitrophthalic acid, 3-nitrophtalic acid, m-nitrophthalic acid
PubChem CID 69043
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])C(=O)O)C(=O)O

Specifications

Melting Point 213.0°C to 216.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 09,823
Solubility Information Solubility in water: 20g/L (25 c). Other solubilities: 1g/10mL methanol
Formula Weight 211.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3-Nitrophthalic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Nitrophthalic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.