missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalimide, 98%

Product Code. 11428156
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428156 5 g 5g
11438156 25 g 25g
11448156 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428156 Supplier Thermo Scientific Alfa Aesar Supplier No. A15291.06
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 603-62-3 | C8H4N2O4 | 192.13 g/mol

3-Nitrophthalimide is a useful dye for biological research purposes. It is a nitro heterocyclic compounds found to exhibit potential antifungal activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrophthalimide is a useful dye for biological research purposes. It is a nitro heterocyclic compounds found to exhibit potential antifungal activities.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 603-62-3
Molecular Formula C8H4N2O4
Molecular Weight (g/mol) 192.13
MDL Number MFCD00041852
InChI Key BONIIQYTWOPUQI-UHFFFAOYSA-N
Synonym 3-nitrophthalimide, 4-nitroisoindoline-1,3-dione, phthalimide, 3-nitro, 1h-isoindole-1,3 2h-dione, 4-nitro, nitrophthalimide, 4-nitro-2,3-dihydro-1h-isoindole-1,3-dione, 4-nitro-1h-isoindole-1,3 2h-dione, 4-nitro-2h-benzo c azoline-1,3-dione, 3-nitro-phthalimide, pubchem12007
PubChem CID 11779
IUPAC Name 4-nitroisoindole-1,3-dione
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])C(=O)NC2=O

Especificaciones

Melting Point 217°C to 221°C
Quantity 5 g
Beilstein 179965
Solubility Information Insoluble in water.
Formula Weight 192.13
Percent Purity 98%
Chemical Name or Material 3-Nitrophthalimide

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.