missing translation for 'onlineSavingsMsg'
Learn More

3-O-Allyl Rocuronium Bromide (>90%), TRC

Product Code. 30422250
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422250 25 mg 25mg
30415500 10 mg 10mg
30424627 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30422250 Supplier Toronto Research Chemicals Supplier No. TRCP76955025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-O-Allyl Rocuronium Bromide
Analyte or Component Names 3-O-Allyl Rocuronium Bromide
Shipping Condition Room Temperature
Molecular Formula C35 H57 N2 O4 . Br
Synonym 3-Propenoxy Rocuronium Bromide, Rocuronium Bromide Impurity 1
InChI Formula InChI=1S/C35H57N2O4.BrH/c1-6-16-37(17-8-9-18-37)31-23-29-27-11-10-26-22-32(40-19-7-2)30(36-14-20-39-21-15-36)24-35(26,5)28(27)12-13-34(29,4)33(31)41-25(3)38;/h6-7,26-33H,1-2,8-24H2,3-5H3;1H/q+1;/p-1/t26-,27+,28-,29-,30-,31-,32-,33-,34-,35-;/m0./s1
Solution Type Neat
SMILES [Br-].CC(=O)O[C@H]1[C@H](C[C@H]2[C@@H]3CC[C@H]4C[C@H](OCC=C)[C@H](C[C@]4(C)[C@H]3CC[C@]12C)N5CCOCC5)[N+]6(CC=C)CCCC6
IUPAC Name [(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-3-allyloxy-16-(1-allylpyrrolidin-1-ium-1-yl)-10,13-dimethyl-2-morpholino-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate;bromide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 649.74
Quantity 25 mg
Formula Weight 648.3502
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.