missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxy-1-propanol, 97%

Product Code. 10032280 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10032280

Brand: Thermo Scientific Acros 186980050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6180-61-6 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6180-61-6
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00039549
InChI Key AWVDYRFLCAZENH-UHFFFAOYSA-N
Synonym 3-phenoxy-1-propanol, 1-propanol, 3-phenoxy, 3-phenoxy 1-propanol, unii-515pbp1u72, propoxylated phenol, ph 4p, polypropylene glycol phenyl ether, 3-phenoxypropanol, 3-phenyloxypropanol, polypropylene glycol monophenyl ether
PubChem CID 80319
IUPAC Name 3-phenoxypropan-1-ol
SMILES OCCCOC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.0400g/mL
Boiling Point 83°C to 85°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5O(CH2)3OH
Refractive Index 1.5280 to 1.53
Quantity 5 g
Beilstein 06, 147
Specific Gravity 1.04
Formula Weight 152.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Phenoxy-1-propanol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Phenoxy-1-propanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.