missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxyanisole, 97%

Product Code. 15465207
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465207 25 g 25g
15455207 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465207 Supplier Thermo Scientific Alfa Aesar Supplier No. H32482.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1655-68-1 | C13H12O2 | 200.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1655-68-1
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
MDL Number MFCD00455605
InChI Key CBVXNDCIOLXDFD-UHFFFAOYSA-N
PubChem CID 74252
IUPAC Name 1-methoxy-3-phenoxybenzene
SMILES COC1=CC=CC(OC2=CC=CC=C2)=C1

Specifications

Odor Odorless
Refractive Index 1.579
Quantity 25 g
Synonym benzene, 1-methoxy-3-phenoxy, m-methoxyphenyl phenyl ether, m-phenoxyanisole, 3-phenoxyanisole, m-phenoxymethoxybenzene, 3-methoxyphenoxybenzene, m-phenoxyphenol monomethyl ether, 3-methoxydiphenyl ether, benzene,1-methoxy-3-phenoxy, benzene, 1-methoxy-3-phenyloxy
Solubility Information Insoluble in water.
Formula Weight 200.23
Percent Purity 97%
Chemical Name or Material 3-Phenoxyanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.