missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzeneboronic acid, 97+%

Product Code. 15495637
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495637 250 mg 250mg
15485637 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495637 Supplier Thermo Scientific Alfa Aesar Supplier No. H32879.MD
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 221006-66-2 | C12H11BO3 | 214.03 g/mol

The extensive use of products containing PBDEs has resulted in the release of 3-Phenoxybenzeneboronic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The extensive use of products containing PBDEs has resulted in the release of 3-Phenoxybenzeneboronic acid.

Solubility
Slightly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 221006-66-2
Molecular Formula C12H11BO3
Molecular Weight (g/mol) 214.03
MDL Number MFCD07780272
InChI Key LOPQWMNOCSRRSR-UHFFFAOYSA-N
Synonym 3-phenoxyphenyl boronic acid, 3-phenoxybenzeneboronic acid, boronic acid, 3-phenoxyphenyl, boronic acid, b-3-phenoxyphenyl, acmc-1cmad
PubChem CID 10130574
IUPAC Name (3-phenoxyphenyl)boronic acid
SMILES OB(O)C1=CC(OC2=CC=CC=C2)=CC=C1

Specifications

Melting Point 138°C to 143°C
Quantity 250 mg
Solubility Information Slightly soluble.
Formula Weight 214.03
Percent Purity ≥97%
Chemical Name or Material 3-Phenoxybenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.