missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzyl alcohol, 98%

Product Code. 11429913
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429913 250 g 250g
11409913 10 g 10g
11419913 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429913 Supplier Thermo Scientific Alfa Aesar Supplier No. A13008.30
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13826-35-2 | C13H12O2 | 200.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-35-2
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00004636
InChI Key KGANAERDZBAECK-UHFFFAOYSA-N
Synonym 3-phenoxybenzyl alcohol, 3-phenoxyphenyl methanol, 3-phenoxybenzylalcohol, m-phenoxybenzyl alcohol, 3-phenoxybenzenemethanol, benzenemethanol, 3-phenoxy, 3-hydroxymethyl diphenyl ether, 3-phenoxybenzylic alcohol, benzyl alcohol, m-phenoxy, 3-pboh
PubChem CID 26295
ChEBI CHEBI:62527
IUPAC Name (3-phenoxyphenyl)methanol
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)CO

Specifications

Melting Point 4°C to 7°C
Density 1.149
Boiling Point 300°C
Flash Point 186°C (366°F)
Refractive Index 1.593
Quantity 250 g
Beilstein 475312
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material 3-Phenoxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.