missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzyl chloride, 97%

Product Code. 11407607
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407607 1 g 1g
11417607 5 g 5g
11427607 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11407607 Supplier Thermo Scientific Alfa Aesar Supplier No. A15670.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 53874-66-1 | C13H11ClO | 218.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53874-66-1
Molecular Formula C13H11ClO
Molecular Weight (g/mol) 218.68
MDL Number MFCD00040866
InChI Key QUYVTGFWFHQVRO-UHFFFAOYSA-N
Synonym 3-phenoxybenzyl chloride, 1-chloromethyl-3-phenoxybenzene, m-phenoxybenzyl chloride, benzene, 1-chloromethyl-3-phenoxy, 3-phenoxyphenylmethyl chloride, 3-phenoxybenzylchloride, 3-chloromethyl-1-phenoxybenzene, 1-chlormethyl-3-phenoxybenzol, acmc-1asdb, m-phenoxy-benzyl chloride
PubChem CID 93291
IUPAC Name 1-(chloromethyl)-3-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)CCl

Specifications

Density 1.189
Boiling Point 141°C to 142°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.591
Quantity 1 g
UN Number UN3265
Beilstein 2212101
Formula Weight 218.69
Percent Purity 97%
Chemical Name or Material 3-Phenoxybenzyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.