missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10694775
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10694775 1 g Amber Glass Bottle 1g
10546534 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10694775 Supplier Thermo Scientific Maybridge Supplier No. CC57163DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 376637-85-3
Molecular Formula C13H14ClNO
Molecular Weight (g/mol) 235.711
MDL Number MFCD07781045
InChI Key WMFHUUKYIUOHRA-UHFFFAOYSA-N
Synonym 3-phenoxybenzylamine hydrochloride, 3-phenoxyphenyl methylamine hydrochloride, 3-phenoxy-benzylamine hydrochloride, 3-phenoxyphenyl methylamine hcl, 3-phenoxyphenyl methanamine hydrochloride, 3-phenoxybenzyl amine hydrochloride, 1-3-phenoxyphenyl methanamine hydrochloride, 3-phenoxyphenyl methylamine, chloride, 1-3-phenoxyphenyl methanamine-hydrogen chloride 1/1
PubChem CID 17749849
IUPAC Name (3-phenoxyphenyl)methanamine;hydrochloride
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)CN.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 235.71
Percent Purity 97%
Chemical Name or Material 3-Phenoxybenzylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.