missing translation for 'onlineSavingsMsg'
Learn More

3-(Phenoxymethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10316474
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10316474 250 mg Amber Glass Bottle 250mg
10021534 1 g Amber Glass Bottle 1g
10537664 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10316474 Supplier Thermo Scientific Maybridge Supplier No. CC63601CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 31719-75-2
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
InChI Key URLYREZIPSJJQU-UHFFFAOYSA-N
Synonym 3-phenoxymethyl benzoic acid, benzoic acid,3-phenoxymethyl, 4rg
PubChem CID 3729884
IUPAC Name 3-(phenoxymethyl)benzoic acid
SMILES C1=CC=C(C=C1)OCC2=CC=CC(=C2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 228.25
Percent Purity 97%
Chemical Name or Material 3-(phenoxymethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.