missing translation for 'onlineSavingsMsg'
Learn More

3-phenoxyphenylboronic acid, Thermo Scientific™

Product Code. 10811891
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10811891 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10811891 Supplier Thermo Scientific Maybridge Supplier No. MAY00180DA
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 221006-66-2
Packaging Amber Glass Bottle
Molecular Formula C12H11BO3
MDL Number MFCD07780272
Quantity 1 g
Synonym 3-phenoxyphenyl boronic acid, 3-phenoxybenzeneboronic acid, boronic acid, 3-phenoxyphenyl, boronic acid, b-3-phenoxyphenyl, acmc-1cmad
InChI Key LOPQWMNOCSRRSR-UHFFFAOYSA-N
SMILES OB(O)C1=CC(OC2=CC=CC=C2)=CC=C1
IUPAC Name (3-phenoxyphenyl)boronic acid
Molecular Weight (g/mol) 214.03
PubChem CID 10130574
Formula Weight 214.03
Percent Purity 97%
Chemical Name or Material 3-phenoxyphenylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.