missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propanol, 98%

Product Code. 10591411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10591411 250 g Glass bottle 250g
10143840 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10591411 Proveedor Thermo Scientific Acros N.º de proveedor 161312500
 Restricted Item
Estimated Shipment: 21-09-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 122-97-4 | C9H12O | 136.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 122-97-4
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00002950
InChI Key VAJVDSVGBWFCLW-UHFFFAOYSA-N
Synonym 3-phenyl-1-propanol, benzenepropanol, hydrocinnamyl alcohol, 3-phenylpropyl alcohol, 3-phenylpropanol, 3-benzenepropanol, phenylpropyl alcohol, hydrocinnamic alcohol, 3-phenyl-n-propanol, 3-hydroxypropyl benzene
PubChem CID 31234
IUPAC Name 3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)CCCO

Especificaciones

CAS Min % 97.5
CAS Max % 100.0
Melting Point -18°C
Color Colorless
Density 1.0400g/mL
Boiling Point 235°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5(CH2)3OH
Refractive Index 1.5250 to 1.527
Quantity 250 g
Beilstein 06, 503
Specific Gravity 1.04
Solubility Information Solubility in water: 10.3g/L water (20°C). Other solubilities: soluble in methanol,cyclohexane,benzene and,ethylacetate
Viscosity 20 mPa.s (20°C)
Formula Weight 136.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Phenyl-1-propanol, 98%
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 3-Phenyl-1-propanol
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.