accessibility menu, dialog, popup

UserName

HO

CAS RN 122-97-4

IUPAC Name: 3-phenylpropan-1-ol
Synonyms: phenylpropanol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: VAJVDSVGBWFCLW-UHFFFAOYSA-N
SMILES: OCCCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)