missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propanol, 99%

Product Code. 11499933
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499933 1000 g 1000g
11409943 250 g 250g
11419943 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499933 Supplier Thermo Scientific Alfa Aesar Supplier No. A13022.0B
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 122-97-4 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
3-Phenyl-1-propanol has a natural fragrance with antimicrobial properties against bacteria and molds. It is used in combination with Heliotropin or Piperonal as a preservative for cosmetic products. Its ester derivatives are used as fragrance component in fresh flower compositions, like lilac, hyacinth and lily of the valley because of the balsamic odor character. It is also a useful biochemical for proteomics research.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-97-4
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002950
InChI Key VAJVDSVGBWFCLW-UHFFFAOYSA-N
Synonym 3-phenyl-1-propanol, benzenepropanol, hydrocinnamyl alcohol, 3-phenylpropyl alcohol, 3-phenylpropanol, 3-benzenepropanol, phenylpropyl alcohol, hydrocinnamic alcohol, 3-phenyl-n-propanol, 3-hydroxypropyl benzene
PubChem CID 31234
IUPAC Name 3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)CCCO

Specifications

Melting Point -18°C
Density 1.004
Boiling Point 235°C
Flash Point 109°C (228°F)
Linear Formula C6H5(CH2)3OH
Refractive Index 1.526
Quantity 1000 g
Beilstein 1857542
Solubility Information Not miscible in water.
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material 3-Phenyl-1-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.