missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropiophenone, 98%

Product Code. 11327266
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327266 50 g 50g
11317266 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327266 Supplier Thermo Scientific Alfa Aesar Supplier No. L09173.18
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1083-30-3 | C15H14O | 210.276 g/mol

3-Phenylpropiophenone is used to produce 2-bromo-1,3-diphenyl-propan-1-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenylpropiophenone is used to produce 2-bromo-1,3-diphenyl-propan-1-one.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1083-30-3
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00039563
InChI Key QGGZBXOADPVUPN-UHFFFAOYSA-N
Synonym dihydrochalcone, 3-phenylpropiophenone, benzylacetophenone, hydrochalcone, 1,3-diphenyl-1-propanone, beta-phenylpropiophenone, hydrocinnamophenone, 1,3-diphenyl-1-oxopropane, phenethyl phenyl ketone, phenyl phenethyl ketone
PubChem CID 64802
ChEBI CHEBI:71231
IUPAC Name 1,3-diphenylpropan-1-one
SMILES C1=CC=C(C=C1)CCC(=O)C2=CC=CC=C2

Specifications

Melting Point 68°C to 73°C
Quantity 50 g
Beilstein 1910661
Solubility Information Insoluble in water.
Formula Weight 210.28
Percent Purity 98%
Chemical Name or Material 3-Phenylpropiophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.