missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropylamine, 98+%

Product Code. 11381996
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11381996 50 g 50g
11371996 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381996 Supplier Thermo Scientific Alfa Aesar Supplier No. L03023.18
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2038-57-5 | C9H13N | 135.21 g/mol

3-Phenylpropylamine is is a compound used for the synthesis of various pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenylpropylamine is is a compound used for the synthesis of various pharmaceuticals.

Solubility
Soluble in Chloroform, Ethyl Acetate. Not miscible in water.

Notes
Store away from air and under dry inert gas. Air sensitive. Incompatible with acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2038-57-5
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008224
InChI Key LYUQWQRTDLVQGA-UHFFFAOYSA-N
Synonym 3-phenylpropylamine, benzenepropanamine, 1-amino-3-phenylpropane, hydrocinnamylamine, gamma-phenylpropylamine, 3-phenyl-n-propylamine, 3-phenyl-1-propylamine, 3-phenyl-1-propanamine, propylamine, 3-phenyl, 3-phenylpropanamine
PubChem CID 16259
IUPAC Name 3-phenylpropan-1-amine
SMILES NCCCC1=CC=CC=C1

Specifications

Density 0.949
Boiling Point 220°C to 221°C
Flash Point 90°C (194°F)
Odor Amine-like
Refractive Index 1.525
Quantity 50 g
UN Number UN2735
Beilstein 2205526
Sensitivity Air Sensitive
Solubility Information Soluble in Chloroform,Ethyl Acetate. Not miscible in water.
Formula Weight 135.21
Percent Purity ≥98%
Chemical Name or Material 3-Phenylpropylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.