missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylsulfonamidopyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Product Code. 15411066
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15411066 1 g 1g
15421066 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15411066 Supplier Thermo Scientific Alfa Aesar Supplier No. H54339.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1083326-28-6
Molecular Formula C17H21BN2O4S
Molecular Weight (g/mol) 360.235
MDL Number MFCD13190589
InChI Key UXJVHVXONVGHIL-UHFFFAOYSA-N
Synonym n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-yl benzenesulfonamide, 3-phenylsulfonamidopyridine-5-boronic acid pinacol ester, n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-pyridinyl benzenesulfonamide, n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-yl benzenesulfonamide, n-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-3-yl benzenesulfonamide
PubChem CID 52936632
IUPAC Name N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl]benzenesulfonamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)NS(=O)(=O)C3=CC=CC=C3

Specifications

Quantity 1 g
Formula Weight 360.24
Percent Purity 96%
Chemical Name or Material 3-Phenylsulfonamidopyridine-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.