missing translation for 'onlineSavingsMsg'
Learn More

3-(Phenylsulfonyl)propionitrile, 99%

Product Code. 11389026
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389026 25 g 25g
11379026 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389026 Supplier Thermo Scientific Alfa Aesar Supplier No. L20196.14
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10154-75-3 | C9H9NO2S | 195.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10154-75-3
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.236
MDL Number MFCD00047798
InChI Key OVZPZPVSUAKTCM-UHFFFAOYSA-N
Synonym 3-phenylsulfonyl propionitrile, 3-phenylsulfonyl propanenitrile, 3-phenylsulphonyl propiononitrile, 3-benzenesulfonyl propanenitrile, 3-phenylsulfonylpropionitrile, acmc-1bvrh, enamine_005271, 3-phenylsulfonyl propanitrile, 3-phenylsulphonyl-propionitrile, propanenitrile, 3-phenylsulfonyl
PubChem CID 82413
IUPAC Name 3-(benzenesulfonyl)propanenitrile
SMILES C1=CC=C(C=C1)S(=O)(=O)CCC#N

Specifications

Melting Point 94°C to 96°C
Odor Odorless
Quantity 25 g
Beilstein 2051019
Formula Weight 195.24
Percent Purity 99%
Chemical Name or Material 3-(Phenylsulfonyl)propionitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.