missing translation for 'onlineSavingsMsg'
Learn More

3-(Pyrid-2-yloxy)benzoic acid, 97%, Thermo Scientific™

Product Code. 10788075
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10788075 250 mg Amber Glass Bottle 250mg
10604975 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10788075 Supplier Thermo Scientific Maybridge Supplier No. CC50201CB
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 51362-30-2
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.208
MDL Number MFCD09025853
InChI Key LYSIEAIIZBZRCE-UHFFFAOYSA-N
Synonym 3-pyridin-2-yloxy benzoic acid, 3-pyrid-2-yloxy benzoic acid, 3-2-pyridyloxy benzoic acid, 3-2 pyridyloxy-benzoic acid, 3-pyridin-2-yl oxy benzoic acid, benzoic acid, 3-2-pyridinyloxy
PubChem CID 21901423
IUPAC Name 3-pyridin-2-yloxybenzoic acid
SMILES C1=CC=NC(=C1)OC2=CC=CC(=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 215.21
Percent Purity 95%
Chemical Name or Material 3-(Pyrid-2-yloxy)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.