missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-2-ylbenzoic acid, 97%, Thermo Scientific™

Product Code. 10386954
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10386954 5 g Amber Glass Bottle 5g
10346334 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10386954 Supplier Thermo Scientific Maybridge Supplier No. CC56301DE
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 579476-26-9
Molecular Formula C11H8N2O2
Molecular Weight (g/mol) 200.197
MDL Number MFCD05864812
InChI Key KLTNGIZIUGYITR-UHFFFAOYSA-N
PubChem CID 22317373
IUPAC Name 3-pyrimidin-2-ylbenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C2=NC=CC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 3-pyrimidin-2-yl-benzoic acid, 3-pyrimidin-2-yl benzoic acid, 2-3-carboxyphenyl pyrimidine, 3-2-pyrimidinyl benzoic acid, benzoic acid,3-2-pyrimidinyl, benzoic acid, 3-2-pyrimidinyl, 3-pyrimidin-2-yl benzoicacid
Formula Weight 200.2
Percent Purity 97%
Chemical Name or Material 3-Pyrimidin-2-ylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.