missing translation for 'onlineSavingsMsg'
Learn More

(3-Pyrimidin-2-ylphenyl)methanol, 97%, Thermo Scientific™

Product Code. 10444895
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10444895 1 g Amber Glass Bottle 1g
10462274 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10444895 Supplier Thermo Scientific Maybridge Supplier No. CC56309DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 892502-12-4
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.21
MDL Number MFCD09064977
InChI Key WMNXGJMCXOLBBV-UHFFFAOYSA-N
Synonym 3-pyrimidin-2-ylphenyl methanol, 3-pyrimidin-2-yl phenyl methanol, benzenemethanol,3-2-pyrimidinyl, 3-pyrimidin-2-yl-benzyl alcohol, benzenemethanol, 3-2-pyrimidinyl, 3-pyrimidin-2-ylphenyl methan-1-ol
PubChem CID 24229565
SMILES OCC1=CC=CC(=C1)C1=NC=CC=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name (3-pyrimidin-2-ylphenyl)methanol
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material (3-Pyrimidin-2-ylphenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.