missing translation for 'onlineSavingsMsg'
Learn More

3-Sulfamoylbenzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15409387
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15409387 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15409387 Supplier Thermo Scientific Alfa Aesar Supplier No. H52628.MD
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 850568-74-0
Melting Point 260°C to 266°C
Molecular Formula C6H8BNO4S
MDL Number MFCD06659840
Quantity 250 mg
Synonym 3-boronobenzenesulfonamide, 3-aminosulphonyl benzeneboronic acid, 3-sulfamoylphenyl boronic acid, 3-boronobenzenesulphonamide, boronic acid b-3-aminosulfony pheny, boronic acid, 3-aminosulfonyl phenyl, 3-aminosulfonylphenyl boronic acid, acmc-209q2m
InChI Key PMNJSWJJYWZLHU-UHFFFAOYSA-N
SMILES NS(=O)(=O)C1=CC=CC(=C1)B(O)O
IUPAC Name (3-sulfamoylphenyl)boronic acid
Molecular Weight (g/mol) 201.00
PubChem CID 44119323
Formula Weight 201
Percent Purity 97%
Chemical Name or Material 3-Sulfamoylbenzeneboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.