missing translation for 'onlineSavingsMsg'
Learn More

3-Sulfobenzoic acid monosodium salt, 97%

Product Code. 11425577
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425577 25 g 25g
11435577 100 g 100g
11445577 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425577 Supplier Thermo Scientific Alfa Aesar Supplier No. B23556.14
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 17625-03-5 | C7H5NaO5S | 224.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17625-03-5
Molecular Formula C7H5NaO5S
Molecular Weight (g/mol) 224.162
MDL Number MFCD00066378
InChI Key KQHKITXZJDOIOD-UHFFFAOYSA-M
PubChem CID 23668512
IUPAC Name sodium;3-sulfobenzoate
SMILES C1=CC(=CC(=C1)S(=O)(=O)O)C(=O)[O-].[Na+]

Specifications

Quantity 25 g
Sensitivity Hygroscopic
Synonym sodium 3-sulfobenzoate, 3-sulfobenzoic acid monosodium salt, benzoic acid, 3-sulfo-, monosodium salt, benzoic acid, 3-sulfo-, sodium salt 1:1, sodium hydrogen m-sulphonatobenzoate, sodium3-sulfobenzoate, sodium m-sulfobenzoate, acmc-209sh5, 3-sodiosulfo benzoic acid, ksc491e3l
Formula Weight 224.17
Percent Purity 97%
Chemical Name or Material 3-Sulfobenzoic acid monosodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.