missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butyldimethylsilyloxy)phenylboronic acid, 96+%, Thermo Scientific Chemicals

Product Code. 10112374 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10112374 1 g Glass bottle 1g
10011354 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10112374

Brand: Thermo Scientific Acros 398340010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261621-12-9
Molecular Formula C12H21BO3Si
Molecular Weight (g/mol) 252.19
MDL Number MFCD03701505
InChI Key RDQWADDNQONTLB-UHFFFAOYSA-N
Synonym 3-tert-butyldimethylsilyl oxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy phenylboronic acid, 3-t-butyldimethylsilyloxy phenylboronic acid, 3-tert-butyl dimethylsiloxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy benzeneboronic acid, 3-tert-butyldimethylsiloxy benzeneboronic acid, 3-tert-butyldimethylsilyl oxy phenylboronic acid, 3-tert-butyl dimethyl silyl oxyphenyl boronic acid, m-t-butyldimethylsilyloxy phenylboronic acid, 3-t-butyl dimethylsiloxy phenyl boronic acid
PubChem CID 3463029
IUPAC Name [3-[tert-butyl(dimethyl)silyl]oxyphenyl]boronic acid
SMILES CC(C)(C)[Si](C)(C)OC1=CC=CC(=C1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 86°C to 91°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96+%
Packaging Glass bottle
Linear Formula (CH3)CSi(CH3)2OC6H4B(OH)2
Quantity 1 g
Formula Weight 252.19
Percent Purity 96+%
Physical Form Powder
Chemical Name or Material 3-(tert-Butyldimethylsilyloxy)phenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-(tert-Butyldimethylsilyloxy)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.