missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 10419960 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10419960 500 mg Glass Bottle 500mg
10772875 2.5 g Glass Bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10419960 Supplier Thermo Scientific Acros Supplier No. 297635000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1535-73-5
MDL Number MFCD00041511
InChI Key SADHVOSOZBAAGL-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy aniline, 3-trifluoromethoxy-phenylamine, m-aminophenyl trifluoromethyl ether, m-trifluoromethoxy aniline, benzenamine, 3-trifluoromethoxy, alpha,alpha,alpha-trifluoro-m-anisidine, 3-trifluoromethoxy benzenamine, 3-trifluoromethoxy phenylamine, pubchem4507
PubChem CID 73753
IUPAC Name 3-(trifluoromethoxy)aniline
SMILES C1=CC(=CC(=C1)OC(F)(F)F)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Orange
Density 1.3200g/mL
Boiling Point 72.0°C to 75.0°C (8.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C7H6F3NO
Linear Formula CF3OC6H4NH2
Refractive Index 1.4650 to 1.4670
Quantity 500 mg
Specific Gravity 1.32
Molecular Weight (g/mol) 177.12
Formula Weight 177.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethoxy)aniline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Trifluoromethoxy)aniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.