missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzeneboronic acid, 98%

Product Code. 11389725
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389725 1 g 1g
11399725 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389725 Supplier Thermo Scientific Alfa Aesar Supplier No. L19775.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 179113-90-7 | C7H6BF3O3 | 205.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179113-90-7
Molecular Formula C7H6BF3O3
Molecular Weight (g/mol) 205.93
MDL Number MFCD01320697
InChI Key UWDFWVLAHRQSKK-UHFFFAOYSA-N
Synonym 3-trifluoromethoxyphenylboronic acid, 3-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boronic acid, 3-trifluoromethoxy benzeneboronic acid, m-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boranediol, 3-trifluoromethyloxy phenylboronic acid, boronic acid, 3-trifluoromethoxy phenyl
PubChem CID 2734385
IUPAC Name [3-(trifluoromethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=CC(OC(F)(F)F)=C1

Specifications

Melting Point ∼80°C
Quantity 1 g
Beilstein 7531456
Formula Weight 205.93
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethoxy)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.