missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzoic acid, 98%, Thermo Scientific™

Product Code. 15565066 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15565066 25 g 25g
15555066 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15565066 Supplier Thermo Scientific Acros Supplier No. 297660250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1014-81-9
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
InChI Key OKPFIWIMBJNFSE-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzoic acid, m-trifluoromethoxybenzoic acid, m-trifluoromethoxy benzoic acid, benzoic acid, 3-trifluoromethoxy, 3-trifluoromethoxy benzoicacid, alpha,alpha,alpha-trifluoro-m-anisic acid, 3-carboxy-alpha,alpha,alpha-trifluoroanisole, 3-trifluoromethoxy-benzoic acid, pubchem4590, acmc-1btyb
PubChem CID 66096
IUPAC Name 3-(trifluoromethoxy)benzoic acid
SMILES C1=CC(=CC(=C1)OC(F)(F)F)C(=O)O

Specifications

Melting Point 91.0°C to 93.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Linear Formula CF3OC6H4CO2H
Quantity 25 g
Formula Weight 206.12
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethoxy)benzoic acid
compliance-icons
Product Identifier
  • 3-(Trifluoromethoxy)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.