missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzoic acid, 98%, Thermo Scientific™

Product Code. 10553031 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10553031 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10553031 Supplier Thermo Scientific Acros Supplier No. 297660010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1014-81-9
CAS Min % 99.0
CAS Max % 100.0
Melting Point 91.0°C to 93.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C8H5F3O3
Linear Formula CF3OC6H4CO2H
MDL Number MFCD00041501
Quantity 1 g
Synonym 3-trifluoromethoxy benzoic acid, m-trifluoromethoxybenzoic acid, m-trifluoromethoxy benzoic acid, benzoic acid, 3-trifluoromethoxy, 3-trifluoromethoxy benzoicacid, alpha,alpha,alpha-trifluoro-m-anisic acid, 3-carboxy-alpha,alpha,alpha-trifluoroanisole, 3-trifluoromethoxy-benzoic acid, pubchem4590, acmc-1btyb
InChI Key OKPFIWIMBJNFSE-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)OC(F)(F)F)C(=O)O
IUPAC Name 3-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol) 206.12
PubChem CID 66096
Formula Weight 206.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-(Trifluoromethoxy)benzoic acid, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Trifluoromethoxy)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.