missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzyl bromide, 98%

Product Code. 11426047
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426047 5 g 5g
11416047 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11426047 Supplier Thermo Scientific Alfa Aesar Supplier No. B25061.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 159689-88-0 | C8H6BrF3O | 255.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159689-88-0
Molecular Formula C8H6BrF3O
Molecular Weight (g/mol) 255.03
MDL Number MFCD00061271
InChI Key QSIVWRRHVXSDNE-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzyl bromide, 1-bromomethyl-3-trifluoromethoxy benzene, 3-trifluoromethoxybenzyl bromide, alpha-bromo-3-trifluoromethoxy toluene, 3-trifluoromethoxy benzylbromide, m-trifluoromethoxy benzyl bromide, 3-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-3-trifluoromethoxy-benzene, benzene, 1-bromomethyl-3-trifluoromethoxy
PubChem CID 737176
IUPAC Name 1-(bromomethyl)-3-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=CC(CBr)=C1

Specifications

Density 1.572
Boiling Point 178°C to 180°C
Flash Point 88°C (190°F)
Refractive Index 1.478
Quantity 5 g
UN Number UN3265
Beilstein 8052203
Formula Weight 255.04
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethoxy)benzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.