missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)phenol, 98%, Thermo Scientific™

Product Code. 10183424 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10183424

Brand: Thermo Scientific Acros 308702500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 827-99-6
Molecular Formula C7H5F3O2
Molecular Weight (g/mol) 178.11
MDL Number MFCD00040987
InChI Key UWLJERQTLRORJN-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy phenol, m-trifluoromethoxy phenol, phenol, 3-trifluoromethoxy, 3-trifluoromethoxy-phenol, 3-hydroxyphenyl trifluoromethyl ether, pubchem1505, m-trifluoromethoxyphenol, pubchem10391
PubChem CID 2733261
IUPAC Name 3-(trifluoromethoxy)phenol
SMILES OC1=CC=CC(OC(F)(F)F)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3700g/mL
Boiling Point 69.0°C to 70.0°C (12.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula CF3OC6H4OH
Refractive Index 1.4450 to 1.4470
Quantity 250 mg
Specific Gravity 1.37
Formula Weight 178.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethoxy)phenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.