missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)thiophenol, 98%

Product Code. 11349945
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349945 1 g 1g
11359945 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349945 Supplier Thermo Scientific Alfa Aesar Supplier No. L19912.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 220239-66-7 | C7H5F3OS | 194.171 g/mol

3-(Trifluoromethoxy)thiophenol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trifluoromethoxy)thiophenol is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220239-66-7
Molecular Formula C7H5F3OS
Molecular Weight (g/mol) 194.171
MDL Number MFCD01320799
InChI Key GEJGGOYNWFQKKH-UHFFFAOYSA-N
PubChem CID 2777357
IUPAC Name 3-(trifluoromethoxy)benzenethiol
SMILES C1=CC(=CC(=C1)S)OC(F)(F)F

Specifications

Refractive Index 1.477
Quantity 1 g
UN Number UN2810
Sensitivity Air Sensitive
Synonym 3-trifluoromethoxy thiophenol, 3-trifluoromethoxy benzenethiol, 3-trifluoromethoxy benzene-1-thiol, benzenethiol, 3-trifluoromethoxy, acmc-1cejb, 3-trifluoromethoxythiophenol, 3-trifluoromethoxy benzene thiol, 3-trifluoromethyloxy benzenethiol, benzenethiol,3-trifluoromethoxy, 3-thio-alpha,alpha,alpha-trifluoroanisole
Solubility Information Not miscible in water.
Formula Weight 194.18
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethoxy)thiophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.