missing translation for 'onlineSavingsMsg'
Learn More

3-Trifluoromethoxyphenylboronic acid, 98%, Thermo Scientific™

Product Code. 10170462 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10170462 1 g 1g
10320334 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10170462 Supplier Thermo Scientific Acros Supplier No. 359720010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 179113-90-7
Molecular Formula C7H6BF3O3
Molecular Weight (g/mol) 205.93
MDL Number MFCD01320697
InChI Key UWDFWVLAHRQSKK-UHFFFAOYSA-N
Synonym 3-trifluoromethoxyphenylboronic acid, 3-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boronic acid, 3-trifluoromethoxy benzeneboronic acid, m-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boranediol, 3-trifluoromethyloxy phenylboronic acid, boronic acid, 3-trifluoromethoxy phenyl
PubChem CID 2734385
IUPAC Name [3-(trifluoromethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=CC(OC(F)(F)F)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 79°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 205.93
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 3-Trifluoromethoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Trifluoromethoxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.