missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)aniline, 99%

Product Code. 11478436
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478436 100 g 100g
11488436 500 g 500g
11498436 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478436 Supplier Thermo Scientific Alfa Aesar Supplier No. A15910.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 98-16-8 | C7H6F3N | 161.127 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Soluble in water. [5 g/L (20°C)]

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-16-8
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.127
MDL Number MFCD00007797
InChI Key VIUDTWATMPPKEL-UHFFFAOYSA-N
Synonym 3-trifluoromethyl aniline, 3-aminobenzotrifluoride, m-aminobenzotrifluoride, 3-aminotrifluorotoluene, benzenamine, 3-trifluoromethyl, 3-trifluoromethyl benzenamine, m-abtf, m-trifluoromethylaniline, m-amino trifluorotoluene, m-trifluoromethyl aniline
PubChem CID 7375
IUPAC Name 3-(trifluoromethyl)aniline
SMILES C1=CC(=CC(=C1)N)C(F)(F)F

Specifications

Melting Point 5°C to 6°C
Density 1.295
Boiling Point 187°C to 189°C
Flash Point 85°C (185°F)
Odor Recognizable
Refractive Index 1.48
Quantity 100 g
UN Number UN2948
Beilstein 387672
Solubility Information Soluble in water. [5g/L (20°C)]
Formula Weight 161.13
Percent Purity 99%
Chemical Name or Material 3-(Trifluoromethyl)aniline
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-(Trifluoromethyl)aniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 3-(Trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.