missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzenesulfonyl chloride, 95%, Thermo Scientific™

Product Code. 10751151 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10751151 5 g Glass bottle 5g
10743465 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10751151 Supplier Thermo Scientific Acros Supplier No. 306280050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 777-44-6
Molecular Formula C7H4ClF3O2S
Molecular Weight (g/mol) 244.62
MDL Number MFCD00014724
InChI Key ONCAZCNPWWQQMW-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzenesulfonyl chloride, 3-trifluoromethyl benzene-1-sulfonyl chloride, m-trifluoromethylbenzenesulfonyl chloride, 3-trifluoromethyl benzenesulphonyl chloride, 3-trifluoromethylbenzenesulfochloride, 3-trifluoromethyl benzenesulfonylchloride, benzenesulfonyl chloride, 3-trifluoromethyl, 3-chlorosulphonyl benzotrifluoride, m-trifluoromethylphenylsulfonyl chloride, 3-trifluoromethyl-benzenesulfonyl chloride
PubChem CID 2733250
IUPAC Name 3-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)C(F)(F)F

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.5200g/mL
Boiling Point 77°C to 79°C (0.8 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H4SO2Cl
Refractive Index 1.4847 to 1.4867
Quantity 5 g
Specific Gravity 1.52
Solubility Information Solubility in water: reacts
Formula Weight 244.62
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)benzenesulfonyl chloride, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.