missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzyl alcohol, 97%

Product Code. 11412758
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412758 25 g 25g
11402758 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11412758 Supplier Thermo Scientific Alfa Aesar Supplier No. A16739.14
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 349-75-7 | C8H7F3O | 176.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349-75-7
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.138
MDL Number MFCD00004645
InChI Key BXEHKCUWIODEDE-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl alcohol, 3-trifluoromethylbenzyl alcohol, 3-trifluoromethyl phenyl methanol, 3-trifluoromethyl benzylalcohol, m-trifluoromethylbenzyl alcohol, m-trifluoromethyl benzyl alcohol, 3-trifluoromethyl benzylic alcohol, benzenemethanol, 3-trifluoromethyl, m-trifluorome benzyl alcohol
PubChem CID 67681
IUPAC Name [3-(trifluoromethyl)phenyl]methanol
SMILES C1=CC(=CC(=C1)C(F)(F)F)CO

Specifications

Density 1.293
Boiling Point 79°C to 80°C (3 mmHg)
Flash Point 84°C (183°F)
Refractive Index 1.46
Quantity 25 g
Beilstein 1450161
Formula Weight 176.14
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)benzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.