missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzyl chloride, 98%

Product Code. 11427717
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427717 5 g 5g
11437717 25 g 25g
11447717 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427717 Supplier Thermo Scientific Alfa Aesar Supplier No. A16253.06
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 705-29-3 | C8H6ClF3 | 194.581 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 705-29-3
Molecular Formula C8H6ClF3
Molecular Weight (g/mol) 194.581
MDL Number MFCD00000908
InChI Key XGASTRVQNVVYIZ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl chloride, 1-chloromethyl-3-trifluoromethyl benzene, 3-trifluoromethylbenzyl chloride, 3-chloromethyl benzotrifluoride, m-trifluoromethylbenzyl chloride, 3-chloromethyl-benzotrifluoride, benzene, 1-chloromethyl-3-trifluoromethyl, alpha'-chloro-alpha,alpha,alpha-trifluoro-m-xylene, 3-chloromethylbenzotrifluoride, 3-trifluoromethylbenzylchloride
PubChem CID 69716
IUPAC Name 1-(chloromethyl)-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)C(F)(F)F)CCl

Specifications

Density 1.254
Boiling Point 68°C to 70°C (12 mmHg)
Flash Point 74°C (165°F)
Odor Pungent
Refractive Index 1.4645
Quantity 5 g
UN Number UN3265
Beilstein 743242
Formula Weight 194.59
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.