missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzylamine, 98%

Product Code. 10572661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10572661 5 g Glass bottle 5g
10239463 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10572661 Supplier Thermo Scientific Acros Supplier No. 312960050
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2740-83-2 | C8H8F3N | 175.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2740-83-2
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.15
MDL Number MFCD00008117
InChI Key YKNZTUQUXUXTLE-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzylamine, 3-trifluoromethyl phenyl methanamine, 3-trifluoromethylbenzylamine, m-trifluoromethylbenzylamine, m-trifluoromethylbenzyl amine, 3-trifluoromethyl benzyl amine, benzenemethanamine, 3-trifluoromethyl, 3-trifluoromethyl-benzylamine, m-trifluoromethyl benzylamine
PubChem CID 75962
IUPAC Name [3-(trifluoromethyl)phenyl]methanamine
SMILES C1=CC(=CC(=C1)C(F)(F)F)CN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2200g/mL
Boiling Point 93.0°C to 97.0°C (22.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H4CH2NH2
Refractive Index 1.4620 to 1.4640
Quantity 5 g
Specific Gravity 1.22
Solubility Information Solubility in water: insoluble
Formula Weight 175.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)benzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.