missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenol, 98+%

Product Code. 11485123
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485123 5 g 5g
11495123 25 g 25g
11405133 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485123 Supplier Thermo Scientific Alfa Aesar Supplier No. A12784.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 98-17-9 | C7H5F3O | 162.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-17-9
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.111
MDL Number MFCD00002299
InChI Key UGEJOEBBMPOJMT-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenol, 3-hydroxybenzotrifluoride, m-trifluoromethylphenol, m-hydroxybenzotrifluoride, phenol, 3-trifluoromethyl, m-trifluoromethyl phenol, alpha,alpha,alpha-trifluoro-m-cresol, a,a,a-trifluoro-m-cresol
PubChem CID 7376
IUPAC Name 3-(trifluoromethyl)phenol
SMILES C1=CC(=CC(=C1)O)C(F)(F)F

Specifications

Melting Point -2°C
Density 1.336
Boiling Point 177°C to 178°C
Flash Point 74°C (165°F)
Odor Phenol-like
Refractive Index 1.458
Quantity 5 g
UN Number UN3265
Beilstein 2045663
Formula Weight 162.11
Percent Purity ≥98%
Chemical Name or Material 3-(Trifluoromethyl)phenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.