missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenylacetone, 97%, Thermo Scientific™

Product Code. 10741271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10741271 10 g Glass bottle 10g
10425295 1 g Glass bottle 1g
10626164 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10741271 Supplier Thermo Scientific Acros Supplier No. 186160100
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 21906-39-8
Molecular Formula C10H9F3O
Molecular Weight (g/mol) 202.176
MDL Number MFCD00000397
InChI Key JPHQCDCEBDRIOL-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylacetone, 1-3-trifluoromethyl phenyl propan-2-one, m-trifluoromethyl phenylacetone, 1-3-trifluoromethyl phenyl acetone, 3-trifluormethyl phenylacetone, 1-3-trifluoromethyl phenyl-2-propanone, m-trifluoromethylphenylacetone, 2-propanone, 1-3-trifluoromethyl phenyl
PubChem CID 89101
IUPAC Name 1-[3-(trifluoromethyl)phenyl]propan-2-one
SMILES CC(=O)CC1=CC(=CC=C1)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.204g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CF3C6H4CH2COCH3
Refractive Index 1.4560 to 1.4580 (20°C, 589nm)
Quantity 10 g
Formula Weight 202.18
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)phenylacetone
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Trifluoromethyl)phenylacetone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.